Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 5 MG
SDP

Catalog No. 5000364399
Encompass_Preferred

(Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 5 MG

Catalog No. 50-003-64399 Supplier Medchemexpress LLC Supplier No. 5MGHY113128
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-sn-Glycerol 3-phosphate | 57-03-4 | 35.0% | 172.07 | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.