Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-sn-glycerol 3-phosphate (DL-α-glycerol phosphate) | 57-03-4 | MFCD00236373 | 35.0% | 172.07 g/mol | C3H9O6P | 5 MG
SDP

Catalog No. 5000264284
Encompass_Preferred

(Rac)-sn-Glycerol 3-phosphate is the racemic form of sn-glycerol-3-phosphate, a phosphorylated glycerol metabolite used in biochemical and metabolic research. Supplied as a research-grade reagent for in vitro assays, enzymology, and metabolic pathway studies; not for human or clinical use.

  • CAS number 57-03-4
  • Molecular formula C3H9O6P
  • Molecular weight 172.07 g/mol
  • Purity 35.0%
  • Physical appearance colorless to light yellow liquid
  • Solubility in water 175 mg/mL (may require ultrasonic)
  • Storage 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month)
  • Intended for research use only; not for clinical use

Catalog No. 50-002-64284 Supplier Medchemexpress LLC Supplier No. HY113128C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-sn-Glycerol 3-phosphate is the racemic form of sn-glycerol-3-phosphate, a phosphorylated glycerol metabolite used in biochemical and metabolic research. Supplied as a research-grade reagent for in vitro assays, enzymology, and metabolic pathway studies; not for human or clinical use.

  • CAS number 57-03-4
  • Molecular formula C3H9O6P
  • Molecular weight 172.07 g/mol
  • Purity 35.0%
  • Physical appearance colorless to light yellow liquid
  • Solubility in water 175 mg/mL (may require ultrasonic)
  • Storage 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month)
  • Intended for research use only; not for clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.