Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-sphingosylphosphorylcholine | 82970-80-7 | 98.0% | 464.62 g/mol | C23H49N2O5P | 10 MG
SDP

Catalog No. 5000289239
Encompass_Preferred

Rac-sphingosylphosphorylcholine is the racemic form of sphingosylphosphorylcholine, a bioactive sphingolipid used in biochemical and cellular research. Supplied as a solid with defined purity and storage recommendations, it is intended for research applications studying sphingolipid signaling and related pathways.

  • Racemic sphingolipid suitable for biochemical and cellular studies.
  • Purity: 98% as supplied.
  • White to off-white solid appearance.
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Available in small research pack sizes, including a 10 mg vial.
  • Molecular weight 464.62 g/mol; formula C23H49N2O5P.

Catalog No. 50-002-89239 Supplier Medchemexpress LLC Supplier No. HY16513510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rac-sphingosylphosphorylcholine is the racemic form of sphingosylphosphorylcholine, a bioactive sphingolipid used in biochemical and cellular research. Supplied as a solid with defined purity and storage recommendations, it is intended for research applications studying sphingolipid signaling and related pathways.

  • Racemic sphingolipid suitable for biochemical and cellular studies.
  • Purity: 98% as supplied.
  • White to off-white solid appearance.
  • Storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Available in small research pack sizes, including a 10 mg vial.
  • Molecular weight 464.62 g/mol; formula C23H49N2O5P.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.