Learn More
Medchemexpress LLC (Rac)-TBAJ-876 | 2131784-39-7 | 99.9% | 100 MG
SDP
Catalog No. 5000338228
Encompass_Preferred
(Rac)-TBAJ-876 is a racemate of TBAJ-876. It is an inhibitor of *mycobacterium tuberculosis* and an analogue of the anti-tuberculosis agent Bedaquiline. It has potential for tuberculosis research.
  • Racemate of TBAJ-876
  • Inhibitor of mycobacterium tuberculosis
  • Analogue of the anti-tuberculosis agent Bedaquiline
  • Potential for tuberculosis research
Catalog No. 50-003-38228 Supplier Medchemexpress LLC Supplier No. HY128866A100MG
(Rac)-TBAJ-876 is a racemate of TBAJ-876. It is an inhibitor of *mycobacterium tuberculosis* and an analogue of the anti-tuberculosis agent Bedaquiline. It has potential for tuberculosis research.
  • Racemate of TBAJ-876
  • Inhibitor of mycobacterium tuberculosis
  • Analogue of the anti-tuberculosis agent Bedaquiline
  • Potential for tuberculosis research