Learn More
Medchemexpress LLC (Rac)-Tenofovir alafenamide-d5 ((rac)-GS-7340-d5) | 379270-36-7 | 481.50 | 5 MG
SDP
Catalog No. 5000445927
Encompass_Preferred
(Rac)-Tenofovir alafenamide-d5 is a deuterium labeled compound that can be utilized as a tracer and an internal standard for various quantitative analyses.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-45927 Supplier Medchemexpress LLC Supplier No. HYW004356S5MG
(Rac)-Tenofovir alafenamide-d5 is a deuterium labeled compound that can be utilized as a tracer and an internal standard for various quantitative analyses.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs