Learn More
Medchemexpress LLC (Rac)-TTA-P2 | 918430-49-6 | 98.7% | C21H29Cl2FN2O2 | 5 MG
SDP
Catalog No. 5000358187
Encompass_Preferred
(Rac)-TTA-P2 is an isomer of TTA-P2 and can be utilized as an experimental control. TTA-P2 is a potent inhibitor of the T-Type calcium channel, effectively penetrating the CNS and blocking native T-type currents in deep cerebellar nuclear neurons. It completely abolishes the window current for both wild-type and mutant Cav3.1 channels, making it suitable for research related to neurological diseases.
  • Isomer of TTA-P2
  • Experimental control
  • Potent inhibitor of T-Type calcium channel
  • Penetrates CNS well
  • Blocks native T-type currents in deep cerebellar nuclear neurons
  • Abolishes window current for wild-type and mutant Cav3.1 channels
  • Potential for research in neurology diseases
Catalog No. 50-003-58187 Supplier Medchemexpress LLC Supplier No. HY507075MG
(Rac)-TTA-P2 is an isomer of TTA-P2 and can be utilized as an experimental control. TTA-P2 is a potent inhibitor of the T-Type calcium channel, effectively penetrating the CNS and blocking native T-type currents in deep cerebellar nuclear neurons. It completely abolishes the window current for both wild-type and mutant Cav3.1 channels, making it suitable for research related to neurological diseases.
  • Isomer of TTA-P2
  • Experimental control
  • Potent inhibitor of T-Type calcium channel
  • Penetrates CNS well
  • Blocks native T-type currents in deep cerebellar nuclear neurons
  • Abolishes window current for wild-type and mutant Cav3.1 channels
  • Potential for research in neurology diseases