Learn More
Medchemexpress LLC (Rac)-TTA-P2 | 918430-49-6 | MFCD11977740 | 98.7% | 431.37 | C21H29Cl2FN2O2 | 10 MG
SDP
Catalog No. 5000298494
Encompass_Preferred
Racemic TTA-P2 is a research-grade T-type calcium channel inhibitor used in preclinical studies to probe neuronal excitability, pain mechanisms, and epilepsy models. It is supplied as a solid with high purity and documented physicochemical properties for laboratory use.
  • Potent inhibitor of T-type calcium channels for electrophysiology studies.
  • High purity suitable for research applications.
  • Available in small-scale quantities for preclinical experiments.
  • Documented molecular weight and chemical formula for assay planning.
  • Stable solid form convenient for storage and handling.
Catalog No. 50-002-98494 Supplier Medchemexpress LLC Supplier No. HY5070710MG
Racemic TTA-P2 is a research-grade T-type calcium channel inhibitor used in preclinical studies to probe neuronal excitability, pain mechanisms, and epilepsy models. It is supplied as a solid with high purity and documented physicochemical properties for laboratory use.
  • Potent inhibitor of T-type calcium channels for electrophysiology studies.
  • High purity suitable for research applications.
  • Available in small-scale quantities for preclinical experiments.
  • Documented molecular weight and chemical formula for assay planning.
  • Stable solid form convenient for storage and handling.