Learn More
Medchemexpress LLC HY-103138A 5mg Medchemexpress, (Rac)-WAY-161503 CAS:75704-24-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103138A5MG
Encompass_Preferred
Medchemexpress, HY-103138A 5mg (Rac)-WAY-161503 CAS:75704-24-4 (Rac)-WAY-161503 is a potent, selective, highly affinity 5-HT2C receptor agonist with a Ki of 4 nM and an EC50 of 12 nM. (Rac)-WAY-161503 displays higher affinity for 5-HT2C than 5-HT2A and 5-HT2B receptors. (Rac)-WAY-161503 has anti-obesity and antidepressant effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8042