Learn More
Medchemexpress LLC HY-136298 5mg Medchemexpress, (Rac)-X77 CAS:2144491-78-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1362985MG
Encompass_Preferred
Medchemexpress, HY-136298 5mg (Rac)-X77 CAS:2144491-78-9 (Rac)-X77 is a racemate of X77. X77 is a potent non-covalent inhibitor of the main protease of SARS-CoV-2 (SARS-CoV-2 Mpro). X77 binds to SARS-CoV-2 Mpro with a Kd value of 0.057 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1434