Learn More
Medchemexpress LLC Rac-XL177A | 2417089-73-5 | C48H57ClN8O5 | 10MG
SDP
Catalog No. 5000260586
Encompass_Preferred
(Rac)-XL177A is the racemic isomer of XL177A, a highly potent, selective, and irreversible inhibitor of USP7. It stabilizes p53 and induces cancer-cell death in cellular assays, and is used in biochemical and cell-based studies of deubiquitinase-mediated pathways. The compound is supplied as a solid; molecular formula C48H57ClN8O5 and molecular weight 861.47 g·mol⁻¹.
Catalog No. 50-002-60586 Supplier Medchemexpress LLC Supplier No. HY138794A10MG
(Rac)-XL177A is the racemic isomer of XL177A, a highly potent, selective, and irreversible inhibitor of USP7. It stabilizes p53 and induces cancer-cell death in cellular assays, and is used in biochemical and cell-based studies of deubiquitinase-mediated pathways. The compound is supplied as a solid; molecular formula C48H57ClN8O5 and molecular weight 861.47 g·mol⁻¹.