Learn More
Medchemexpress LLC Rac-zevaquenabant | 1610420-28-4 | 99.2% | 547.98 | C25H21ClF3N5O2S | 5 MG
SDP
Catalog No. 5000223290
Encompass_Preferred
(Rac)-Zevaquenabant is a research small molecule described as a cannabinoid receptor type 1 (CB1R) and inducible nitric oxide synthase (iNOS) antagonist with reported CB1R Ki ~5.7 nM. It is used in preclinical studies, including liver fibrosis research, and is supplied as a solid or as a ready-to-use solution in DMSO.
  • Potent CB1R antagonist with reported Ki of 5.7 nM.
  • Also antagonizes inducible nitric oxide synthase (iNOS), useful for dual-pathway studies.
  • High purity: 99.2%.
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular weight 547.98; formula C25H21ClF3N5O2S.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-23290 Supplier Medchemexpress LLC Supplier No. HY1414115MG
(Rac)-Zevaquenabant is a research small molecule described as a cannabinoid receptor type 1 (CB1R) and inducible nitric oxide synthase (iNOS) antagonist with reported CB1R Ki ~5.7 nM. It is used in preclinical studies, including liver fibrosis research, and is supplied as a solid or as a ready-to-use solution in DMSO.
  • Potent CB1R antagonist with reported Ki of 5.7 nM.
  • Also antagonizes inducible nitric oxide synthase (iNOS), useful for dual-pathway studies.
  • High purity: 99.2%.
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular weight 547.98; formula C25H21ClF3N5O2S.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months).
  • For research use only; not for human or clinical use.