Learn More
Medchemexpress LLC (Rac)-ZLc-002 | 99.0% | 231.25 | 100 MG
SDP
Catalog No. 5000436192
Encompass_Preferred
(Rac)-ZLc-002 is an inhibitor of nNOS interaction with nitric oxide synthase 1 adaptor protein (NOS1AP). It suppresses inflammatory nociception and chemotherapy-induced neuropathic pain and synergizes with Paclitaxel to reduce tumor cell viability.
  • Inhibitor of nNOS/NOS1AP interaction.
  • Suppresses inflammatory nociception and chemotherapy-induced neuropathic pain.
  • Synergizes with Paclitaxel to reduce tumor cell viability.
  • Reduces co-immunoprecipitation of NOS1AP with nNOS in primary cultured cortical neurons.
  • Attenuates mechanical and cold allodynia evoked by Paclitaxel in a mouse model.
  • Reduces formalin-evoked nociceptive behavior in rats.
  • Attenuates chronic mild stress (CMS)‐induced anxiogenic behavior in ICR mice.
  • Reverses behavioral effects of glucocorticoids in mice.
Catalog No. 50-004-36192 Supplier Medchemexpress LLC Supplier No. HY124996100MG
(Rac)-ZLc-002 is an inhibitor of nNOS interaction with nitric oxide synthase 1 adaptor protein (NOS1AP). It suppresses inflammatory nociception and chemotherapy-induced neuropathic pain and synergizes with Paclitaxel to reduce tumor cell viability.
  • Inhibitor of nNOS/NOS1AP interaction.
  • Suppresses inflammatory nociception and chemotherapy-induced neuropathic pain.
  • Synergizes with Paclitaxel to reduce tumor cell viability.
  • Reduces co-immunoprecipitation of NOS1AP with nNOS in primary cultured cortical neurons.
  • Attenuates mechanical and cold allodynia evoked by Paclitaxel in a mouse model.
  • Reduces formalin-evoked nociceptive behavior in rats.
  • Attenuates chronic mild stress (CMS)‐induced anxiogenic behavior in ICR mice.
  • Reverses behavioral effects of glucocorticoids in mice.