Learn More
Medchemexpress LLC (Rac)-ZLc-002 | 99.0% | 231.25 | 50 MG
SDP
Catalog No. 5000436751
Encompass_Preferred
(Rac)-ZLc-002 is an inhibitor of nNOS interaction with nitric oxide synthase 1 adaptor protein (NOS1AP). It suppresses inflammatory nociception and chemotherapy-induced neuropathic pain. Additionally, it synergizes with Paclitaxel to reduce tumor cell viability.
  • Reduces co-immunoprecipitation of NOS1AP with nNOS immunoprecipitated from primary cultured cortical neurons
  • Synergizes to reduce tumor cell viability
  • Attenuates mechanical and cold allodynia evoked by Paclitaxel
  • Reduces formalin-evoked nociceptive behavior and Fos-like immunoreactivity in the spinal dorsal horn
  • Attenuates chronic mild stress (CMS)‐induced anxiogenic behavior
Catalog No. 50-004-36751 Supplier Medchemexpress LLC Supplier No. HY12499650MG
(Rac)-ZLc-002 is an inhibitor of nNOS interaction with nitric oxide synthase 1 adaptor protein (NOS1AP). It suppresses inflammatory nociception and chemotherapy-induced neuropathic pain. Additionally, it synergizes with Paclitaxel to reduce tumor cell viability.
  • Reduces co-immunoprecipitation of NOS1AP with nNOS immunoprecipitated from primary cultured cortical neurons
  • Synergizes to reduce tumor cell viability
  • Attenuates mechanical and cold allodynia evoked by Paclitaxel
  • Reduces formalin-evoked nociceptive behavior and Fos-like immunoreactivity in the spinal dorsal horn
  • Attenuates chronic mild stress (CMS)‐induced anxiogenic behavior