Learn More
Aviva Systems Biology Corp RAC1 Antibody - middle region (ARP57798_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57798_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAC1. It was validated on Western Blot. RAC1,ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1),MIG5; TC-25; p21-Rac1; MGC111543; RAC1,ras-related C3 botulinum toxin substrate 1,ras-related C3 botulinum toxin substrate 1,ras-like protein TC25,migration-inducing gene 5,cell migration-inducing gene 5 protein,rho family, small GTP binding protein Rac1The immunogen is a synthetic peptide directed towards the middle region of human RAC1. Alternate names: MGC111543, MIG5, TC-25, p21-Rac1, Rac-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-382