Learn More
Enzo Life Sciences Rac1 inhibitor (10mg). CAS: 733767-34-5
SDP
Catalog No. 502013398
Encompass_Preferred
NSC 23766 has been used to investigate the role of Rac1 in such diverse cellular functions as stem cell mobilization, epithelial cell migration, angiogenesis, leukemia cell migration and growth. Alternative name: N6-[2-[[4-(Diethylamino)-1-methylbutyl]amino]-6-methyl-4-pyrimidinyl]-2-methyl-4,6-quinolinediamine trihydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.
Catalog No. 50-201-3398 Supplier Enzo Life Sciences Supplier No. ENZCHM1160010
NSC 23766 has been used to investigate the role of Rac1 in such diverse cellular functions as stem cell mobilization, epithelial cell migration, angiogenesis, leukemia cell migration and growth. Alternative name: N6-[2-[[4-(Diethylamino)-1-methylbutyl]amino]-6-methyl-4-pyrimidinyl]-2-methyl-4,6-quinolinediamine trihydrochloride. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -80°C.