Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac1 protein, human (sf9, GST) | >95.0% | 500 UG
SDP

Catalog No. 5000293319
Encompass_Preferred

Recombinant human RAC1 protein produced in Sf9 insect cells with an N-terminal GST tag. Supplied as a filtered formulation optimized for biochemical and structural assays, the protein shows >95% purity by reducing SDS-PAGE and is available in sizes including 500 μg.

  • Expressed in Sf9 insect cells with N-terminal GST tag.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Predicted molecular weight 47 kDa; observed ~44 kDa on SDS-PAGE.
  • Formulated in 50 mM Tris, 100 mM NaCl, 1 mM GSH, 10% glycerol, pH 8.0.
  • Supplied as a 0.22 μm filtered solution; avoid repeated freeze-thaw cycles.
  • Store at -80°C; stable at -20°C for short-term storage.
  • Suitable for biochemical assays, binding studies, and structural analysis.

Catalog No. 50-002-93319 Supplier Medchemexpress LLC Supplier No. HYP74608500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human RAC1 protein produced in Sf9 insect cells with an N-terminal GST tag. Supplied as a filtered formulation optimized for biochemical and structural assays, the protein shows >95% purity by reducing SDS-PAGE and is available in sizes including 500 μg.

  • Expressed in Sf9 insect cells with N-terminal GST tag.
  • Greater than 95% purity by reducing SDS-PAGE.
  • Predicted molecular weight 47 kDa; observed ~44 kDa on SDS-PAGE.
  • Formulated in 50 mM Tris, 100 mM NaCl, 1 mM GSH, 10% glycerol, pH 8.0.
  • Supplied as a 0.22 μm filtered solution; avoid repeated freeze-thaw cycles.
  • Store at -80°C; stable at -20°C for short-term storage.
  • Suitable for biochemical assays, binding studies, and structural analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.