Learn More
Aviva Systems Biology Corp Rac2 Antibody - C-terminal region (ARP59652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rac2. It was validated on Western Blot. . Alternate names: AI323801, AI452260ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-768