Learn More
Medchemexpress LLC HY-N2064 100mg Medchemexpress, Racanisodamine CAS:17659-49-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2064100MG
Encompass_Preferred
Medchemexpress, HY-N2064 100mg Racanisodamine CAS:17659-49-3 Racanisodamine is one of the racemic isomers of anisodamine, resembles anisodamine in pharmacological effect. Racanisodamine is a non-selective muscarinic antagonist, used as a component of eye drops for myopic control [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3879