Learn More
Apexbio Technology LLC Raceanisodamine 17659-49-3 250mg
SDP
Catalog No. 5000091503
Encompass_Preferred
Raceanisodamine (CAS 17659-49-3) is a naturally occurring tropane alkaloid primarily derived from the roots of Anisodus tanguticus It functions as a weak antagonist of 1-adrenergic receptors which are critical in regulating smooth muscle contraction cardiovascular activity and neurological processes In receptor-binding studies raceanisodamine inhibited the interaction of WB-4101 and clonidine with brain membrane preparations with reported pKi values of 2 63 and 1 61 respectively Additionally raceanisodamine exhibits antioxidant properties and offers cellular protection against free radical-induced damage These characteristics support its use in research focused on receptor pharmacology oxidative stress and neuroprotective mechanisms
Catalog No. 50-000-91503 Supplier Apexbio Technology LLC Supplier No. C5580250
Raceanisodamine (CAS 17659-49-3) is a naturally occurring tropane alkaloid primarily derived from the roots of Anisodus tanguticus It functions as a weak antagonist of 1-adrenergic receptors which are critical in regulating smooth muscle contraction cardiovascular activity and neurological processes In receptor-binding studies raceanisodamine inhibited the interaction of WB-4101 and clonidine with brain membrane preparations with reported pKi values of 2 63 and 1 61 respectively Additionally raceanisodamine exhibits antioxidant properties and offers cellular protection against free radical-induced damage These characteristics support its use in research focused on receptor pharmacology oxidative stress and neuroprotective mechanisms