Learn More
Cayman Chemical Racecadotril, Purity: ≥98%, CAS: 81110-73-8, 1 g
SDP
Catalog No. 502985491
Encompass_Preferred
A prodrug form of thiorphan; reduces or prevents castor oil-induced diarrhea without delaying intestinal transit in rats at 80 or 100 mg/kg, respectively; decreases duration of diarrhea and increases body weight gain in a neonatal gnotobiotic pig model of human rotavirus-induced diarrhea
Catalog No. 50-298-5491 Supplier Cayman Chemical Supplier No. 203901 G
A prodrug form of thiorphan; reduces or prevents castor oil-induced diarrhea without delaying intestinal transit in rats at 80 or 100 mg/kg, respectively; decreases duration of diarrhea and increases body weight gain in a neonatal gnotobiotic pig model of human rotavirus-induced diarrhea