Learn More
Cayman Chemical Racecadotril, Purity: ≥98%, CAS: 81110-73-8, 500 mg
SDP
Catalog No. 502980945
Encompass_Preferred
A prodrug form of thiorphan; reduces or prevents castor oil-induced diarrhea without delaying intestinal transit in rats at 80 or 100 mg/kg, respectively; decreases duration of diarrhea and increases body weight gain in a neonatal gnotobiotic pig model of human rotavirus-induced diarrhea
Catalog No. 50-298-0945 Supplier Cayman Chemical Supplier No. 20390500 MG
A prodrug form of thiorphan; reduces or prevents castor oil-induced diarrhea without delaying intestinal transit in rats at 80 or 100 mg/kg, respectively; decreases duration of diarrhea and increases body weight gain in a neonatal gnotobiotic pig model of human rotavirus-induced diarrhea