Learn More
Apexbio Technology LLC Racecadotril 81110-73-8 1g
SDP
Catalog No. 5000092616
Encompass_Preferred
Racecadotril (CAS 81110-73-8) also known as acetorphan is a small molecule inhibitor of enkephalinase an enzyme responsible for the degradation of endogenous enkephalins By inhibiting enkephalinase with an IC50 of 4 5 M racecadotril increases the availability of enkephalins thereby modulating the activity of enkephalinergic pathways This mechanism is of interest in studies investigating the regulation of opioid-mediated physiological processes including analgesia and intestinal secretion Racecadotril is widely utilized in biomedical research focused on peptide degradation and the pharmacological modulation of endogenous opioid systems
Catalog No. 50-000-92616 Supplier Apexbio Technology LLC Supplier No. B16491000
Racecadotril (CAS 81110-73-8) also known as acetorphan is a small molecule inhibitor of enkephalinase an enzyme responsible for the degradation of endogenous enkephalins By inhibiting enkephalinase with an IC50 of 4 5 M racecadotril increases the availability of enkephalins thereby modulating the activity of enkephalinergic pathways This mechanism is of interest in studies investigating the regulation of opioid-mediated physiological processes including analgesia and intestinal secretion Racecadotril is widely utilized in biomedical research focused on peptide degradation and the pharmacological modulation of endogenous opioid systems