Learn More
Medchemexpress LLC Racecadotril (Acetorphan) | 81110-73-8 | 99.9% | 385.48 | 500 MG
SDP
Catalog No. 5000383964
Encompass_Preferred
Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. It and its active metabolite Thiorphan inhibit purified NEP activity from mouse brain with Kis of 4500 and 6.1 nM, respectively. It acts as an antidiarrheal agent.
  • Neutral endopeptidase (NEP) inhibitor.
  • Inhibits purified NEP activity from mouse brain.
  • Acts as an antidiarrheal agent.
  • For research use only.
  • Solid appearance.
  • White to off-white color.
Catalog No. 50-003-83964 Supplier Medchemexpress LLC Supplier No. HY17399500MG
Racecadotril (Acetorphan) is a neutral endopeptidase (NEP) inhibitor. It and its active metabolite Thiorphan inhibit purified NEP activity from mouse brain with Kis of 4500 and 6.1 nM, respectively. It acts as an antidiarrheal agent.
  • Neutral endopeptidase (NEP) inhibitor.
  • Inhibits purified NEP activity from mouse brain.
  • Acts as an antidiarrheal agent.
  • For research use only.
  • Solid appearance.
  • White to off-white color.