Learn More
Cayman Chemical Racecadotril-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 1246815-11-1, 25 mg
SDP
Catalog No. 502948245
Encompass_Preferred
An internal standard for the quantification of racecadotril by GC- or LC-MS
Catalog No. 50-294-8245 Supplier Cayman Chemical Supplier No. 3389325 MG
An internal standard for the quantification of racecadotril by GC- or LC-MS