Learn More
Strem, An Ascensus Company CAS# 98327-87-8. racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% rac-BINAP. MFCD00010805. 25G
SDP
Catalog No. 5090109870
Encompass
CAS# 98327-87-8. 25G. racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% rac-BINAP. MFCD00010805. Molecular Formula: C44H32P2. Molecular Weight: 622.70. Color/Form: white to light-yellow xtl. Strem#: 15-0433 www.strem.com/catalog/v3/15-0433/
Catalog No. 50-901-09870 Supplier Strem, An Ascensus Company Supplier No. 15043325G
CAS# 98327-87-8. 25G. racemic-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98% rac-BINAP. MFCD00010805. Molecular Formula: C44H32P2. Molecular Weight: 622.70. Color/Form: white to light-yellow xtl. Strem#: 15-0433 www.strem.com/catalog/v3/15-0433/