Learn More
Medchemexpress LLC Racemic 2,2'-bis(diphenylphosphino)-1,1'-binaphthyl | 98327-87-8 | MFCD00010805 | ≥98.0% | 622.67 g/mol | C44H32P2 | 25g
SDP
Catalog No. 5000268969
Encompass_Preferred
(Rac)-BINAP is the isomer of (R)-BINAP (HY-W017757) and can be used as an experimental control (R)-BINAP is a biochemical reagent that can be used as a biological material or organic compound for life science related research
Catalog No. 50-002-68969 Supplier Medchemexpress LLC Supplier No. HYY000325G
(Rac)-BINAP is the isomer of (R)-BINAP (HY-W017757) and can be used as an experimental control (R)-BINAP is a biochemical reagent that can be used as a biological material or organic compound for life science related research