Learn More
Medchemexpress LLC Racemic 2-propyl-4-pentenoic acid | 1575-72-0 | MFCD00869309 | 98.0% | 142.20 g/mol | C8H14O2 | 50MG
SDP
Catalog No. 5000236025
Encompass_Preferred
(±)-2-Propyl-4-pentenoic acid (4-en-VPA) is the racemic form of a known toxic metabolite of valproic acid used as an analytical and research standard for studies of metabolism and developmental toxicity. The material is supplied as a high-purity solid suitable for analytical workflows and toxicology research, and is provided in small pack sizes for laboratory use.
  • High purity suitable for analytical applications.
  • Provided as a solid powder for convenient handling.
  • Relevant for metabolism and neuroteratogenicity studies.
  • Available in small pack sizes for research-scale experiments.
  • Supplied with supplier-specified analytical data and handling recommendations.
Catalog No. 50-002-36025 Supplier Medchemexpress LLC Supplier No. HY12408750MG
Add to Cart
Add to Cart
(±)-2-Propyl-4-pentenoic acid (4-en-VPA) is the racemic form of a known toxic metabolite of valproic acid used as an analytical and research standard for studies of metabolism and developmental toxicity. The material is supplied as a high-purity solid suitable for analytical workflows and toxicology research, and is provided in small pack sizes for laboratory use.
  • High purity suitable for analytical applications.
  • Provided as a solid powder for convenient handling.
  • Relevant for metabolism and neuroteratogenicity studies.
  • Available in small pack sizes for research-scale experiments.
  • Supplied with supplier-specified analytical data and handling recommendations.