Learn More
Medchemexpress LLC Racemic 3-hydroxyphenylglycine | 31932-87-3 | 99.6% | 167.16 g/mol | C8H9NO3 | 5 MG
SDP
Catalog No. 5000211730
Encompass_Preferred
(Rac)-3-Hydroxyphenylglycine is a racemic mixture of the S- and R-enantiomers of 3-hydroxyphenylglycine. The S-enantiomer is reported to act as a potent, selective agonist of the metabotropic glutamate receptor 1 (mGluR1). The material is supplied as an off-white to gray solid for research use and is commonly used in neuroscience and receptor pharmacology studies.
  • Racemic mixture of S- and R-enantiomers.
  • Potent, selective mGluR1 agonist (activity attributed to the S-enantiomer).
  • High purity, suitable for research applications.
  • Available in small, milligram-scale quantities for screening.
  • Solid form; follow low-temperature storage to maintain stability.
Catalog No. 50-002-11730 Supplier Medchemexpress LLC Supplier No. HY1008415MG
(Rac)-3-Hydroxyphenylglycine is a racemic mixture of the S- and R-enantiomers of 3-hydroxyphenylglycine. The S-enantiomer is reported to act as a potent, selective agonist of the metabotropic glutamate receptor 1 (mGluR1). The material is supplied as an off-white to gray solid for research use and is commonly used in neuroscience and receptor pharmacology studies.
  • Racemic mixture of S- and R-enantiomers.
  • Potent, selective mGluR1 agonist (activity attributed to the S-enantiomer).
  • High purity, suitable for research applications.
  • Available in small, milligram-scale quantities for screening.
  • Solid form; follow low-temperature storage to maintain stability.