Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Racemic anabasine-d4 | 1020719-08-7 | MFCD04039400 | >95.0% | 166.26 g/mol | C10H10D4N2 | 10 MG
SDP

Catalog No. 5000284315
Encompass_Preferred

Anabasine-d4 (racemic) is a deuterium-labeled isotope standard of anabasine intended for analytical and biochemical research. It is used as an internal standard or tracer for quantitation and metabolic studies, and is supplied in a small research-scale quantity for laboratory use. Not for human or clinical applications.

  • Deuterium-labeled racemic anabasine
  • Intended as an isotope-labeled internal standard and tracer
  • Research-scale 10 mg quantity suitable for method development
  • Chemical formula C10H10D4N2; molecular weight 166.26 g/mol
  • CAS number 1020719-08-7; purity >95% (HPLC)

Catalog No. 50-002-84315 Supplier Medchemexpress LLC Supplier No. HYW69887110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anabasine-d4 (racemic) is a deuterium-labeled isotope standard of anabasine intended for analytical and biochemical research. It is used as an internal standard or tracer for quantitation and metabolic studies, and is supplied in a small research-scale quantity for laboratory use. Not for human or clinical applications.

  • Deuterium-labeled racemic anabasine
  • Intended as an isotope-labeled internal standard and tracer
  • Research-scale 10 mg quantity suitable for method development
  • Chemical formula C10H10D4N2; molecular weight 166.26 g/mol
  • CAS number 1020719-08-7; purity >95% (HPLC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.