Learn More
Medchemexpress LLC Racemic anatabine | 2743-90-0 | MFCD00078278 | 99.7% | 160.22 | C10H12N2 | 50MG
SDP
Catalog No. 5000235024
Encompass_Preferred
Racemic anatabine is a high-purity reference standard of the tobacco alkaloid anatabine (CAS 2743-90-0) supplied for research and analytical use. It is intended for laboratory applications such as use as a marker of tobacco exposure and for biochemical or neuroscience studies. The material is provided as a solid with recommended cold-storage conditions and is available in multiple pack sizes, including 50 MG.
  • High purity (≈99.7%) for analytical and reference use.
  • Suitable as a marker of tobacco exposure in analytical assays.
  • Molecular formula C10H12N2; molecular weight 160.22.
  • Available in multiple pack sizes, including 50 MG.
  • Store solid at -20°C for long-term stability; follow solvent storage guidance.
  • Intended for research use only; not for human use.
Catalog No. 50-002-35024 Supplier Medchemexpress LLC Supplier No. HY126047A50MG
Racemic anatabine is a high-purity reference standard of the tobacco alkaloid anatabine (CAS 2743-90-0) supplied for research and analytical use. It is intended for laboratory applications such as use as a marker of tobacco exposure and for biochemical or neuroscience studies. The material is provided as a solid with recommended cold-storage conditions and is available in multiple pack sizes, including 50 MG.
  • High purity (≈99.7%) for analytical and reference use.
  • Suitable as a marker of tobacco exposure in analytical assays.
  • Molecular formula C10H12N2; molecular weight 160.22.
  • Available in multiple pack sizes, including 50 MG.
  • Store solid at -20°C for long-term stability; follow solvent storage guidance.
  • Intended for research use only; not for human use.