Learn More
Medchemexpress LLC Racemic AR-13503 (M1 metabolite of AR-13324) | 1254032-16-0 | 99.1% | 321.37 | C19H19N3O2 | 100MG
SDP
Catalog No. 5000238928
Encompass_Preferred
(rac)-AR-13503 is the racemic M1 metabolite of AR-13324 that acts as a ROCK and PKC inhibitor. It has been reported to inhibit angiogenesis, increase retinal pigment epithelium (RPE) permeability, and reduce aberrant neovascularization in preclinical retinal disease models. The compound has formula C19H19N3O2, molecular weight 321.37 g/mol, and reported purity ~99.1%.
  • Acts as a ROCK and PKC inhibitor.
  • Inhibits angiogenesis in preclinical models.
  • Increases retinal pigment epithelium permeability.
  • Reported purity of approximately 99.1%.
  • Available as research-grade solid in multiple package sizes.
Catalog No. 50-002-38928 Supplier Medchemexpress LLC Supplier No. HY125639100MG
(rac)-AR-13503 is the racemic M1 metabolite of AR-13324 that acts as a ROCK and PKC inhibitor. It has been reported to inhibit angiogenesis, increase retinal pigment epithelium (RPE) permeability, and reduce aberrant neovascularization in preclinical retinal disease models. The compound has formula C19H19N3O2, molecular weight 321.37 g/mol, and reported purity ~99.1%.
  • Acts as a ROCK and PKC inhibitor.
  • Inhibits angiogenesis in preclinical models.
  • Increases retinal pigment epithelium permeability.
  • Reported purity of approximately 99.1%.
  • Available as research-grade solid in multiple package sizes.