Learn More
Medchemexpress LLC Racemic BMS-816336 | 1009365-98-3 | 99.9% | 341.45 g/mol | C21H27NO3 | 5 MG
SDP
Catalog No. 5000248324
Encompass_Preferred
Rac-BMS-816336 is the racemic small-molecule inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) used as a research compound in metabolic and glucocorticoid-related studies. It is supplied as a solid, typically soluble in DMSO, and intended for in vitro and preclinical pharmacology work.
  • Potent 11β-HSD1 inhibition in biochemical assays.
  • Suitable for in vitro and preclinical studies.
  • High reported purity for analytical applications.
  • Soluble in DMSO for formulation flexibility.
  • Available in small milligram quantities for research use.
Catalog No. 50-002-48324 Supplier Medchemexpress LLC Supplier No. HY101930A5MG
Rac-BMS-816336 is the racemic small-molecule inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) used as a research compound in metabolic and glucocorticoid-related studies. It is supplied as a solid, typically soluble in DMSO, and intended for in vitro and preclinical pharmacology work.
  • Potent 11β-HSD1 inhibition in biochemical assays.
  • Suitable for in vitro and preclinical studies.
  • High reported purity for analytical applications.
  • Soluble in DMSO for formulation flexibility.
  • Available in small milligram quantities for research use.