Learn More
Medchemexpress LLC Racemic equol | 94105-90-5 | MFCD00016662 | 99.6% | 242.27 g·mol⁻¹ | C15H14O3 | 200 MG
SDP
Catalog No. 5000213659
Encompass_Preferred
Racemic equol is the racemate of equol, a nonsteroidal estrogen receptor ligand and a metabolite of the soy isoflavones daidzin and daidzein. It is commonly used in research on estrogen receptor pharmacology and isoflavone metabolism, with reported EC50 values of 200 nM for ERα and 74 nM for ERβ.
  • Racemic mixture of equol suitable for receptor biology studies.
  • Shows EC50s of 200 nM (ERα) and 74 nM (ERβ).
  • Molecular weight 242.27 g·mol⁻¹ and formula C15H14O3.
  • High purity (≈99.6%) for consistent experimental results.
  • Solid powder form, stable when stored at low temperatures.
  • Available in multiple pack sizes from milligram to gram quantities.
Catalog No. 50-002-13659 Supplier Medchemexpress LLC Supplier No. HY100583A200MG
Racemic equol is the racemate of equol, a nonsteroidal estrogen receptor ligand and a metabolite of the soy isoflavones daidzin and daidzein. It is commonly used in research on estrogen receptor pharmacology and isoflavone metabolism, with reported EC50 values of 200 nM for ERα and 74 nM for ERβ.
  • Racemic mixture of equol suitable for receptor biology studies.
  • Shows EC50s of 200 nM (ERα) and 74 nM (ERβ).
  • Molecular weight 242.27 g·mol⁻¹ and formula C15H14O3.
  • High purity (≈99.6%) for consistent experimental results.
  • Solid powder form, stable when stored at low temperatures.
  • Available in multiple pack sizes from milligram to gram quantities.