Learn More
Medchemexpress LLC Racemic jasmonic acid-d5 | 2750534-78-0 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY122464S21MG
Encompass_Preferred
Racemic jasmonic acid-d5 is the deuterium-labeled (d5) racemate of jasmonic acid used as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is typically supplied as a small solution-based pack and should be stored frozen for stability.
  • Deuterium-labeled internal standard for quantitative assays.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Formula C12H13D5O3; molecular weight 215.30.
  • Supplied as a solution in ethanol; store at -20 °C for up to 2 years.
  • Analytical-grade purity reported on product page (conflicting values: 97.81% and ≥99.0%).
Catalog No. 50-002-31594