Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Racemic lisofylline | 6493-06-7 | MFCD00871850 | 98.6% | 280.33 g/mol | C13H20N4O3 | 50MG
SDP

Catalog No. 5000235027
Encompass_Preferred

Racemic lisofylline is the racemate of lisofylline, an anti-inflammatory research compound and a metabolite-derived analogue of pentoxifylline. It is used in biochemical and cellular studies for its inhibition of phosphatidic acid and free fatty acid generation, and is supplied as a high-purity solid for research applications.

  • Purity 98.6%.
  • Molecular weight 280.33 g/mol.
  • Molecular formula C13H20N4O3.
  • Available in small quantities including 50 mg.
  • Used in studies of inflammation and oxidative tissue injury.
  • Supplied as a solid powder for biochemical assays.

Catalog No. 50-002-35027 Supplier Medchemexpress LLC Supplier No. HY12604250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Racemic lisofylline is the racemate of lisofylline, an anti-inflammatory research compound and a metabolite-derived analogue of pentoxifylline. It is used in biochemical and cellular studies for its inhibition of phosphatidic acid and free fatty acid generation, and is supplied as a high-purity solid for research applications.

  • Purity 98.6%.
  • Molecular weight 280.33 g/mol.
  • Molecular formula C13H20N4O3.
  • Available in small quantities including 50 mg.
  • Used in studies of inflammation and oxidative tissue injury.
  • Supplied as a solid powder for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.