Learn More
Medchemexpress LLC Racemic MEM 1003 | 165187-25-7 | 99.6% | 10 MG
SDP
Catalog No. 5000233807
Encompass_Preferred
(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist (CAS 165187-25-7) used in neuroscience research. The compound has formula C22H25ClN2O5 and molecular weight 432.9 g·mol⁻¹, and is supplied at high purity for research applications.
  • Racemic form of MEM 1003, a dihydropyridine calcium channel antagonist.
  • Potent L-type Ca2+ channel antagonist; useful in neuronal signaling studies.
  • High purity suitable for research use.
  • Available in small-scale quantities (e.g., 5 mg-100 mg) with larger sizes by request.
  • Certificate of analysis, datasheet, and safety data sheet available.
Catalog No. 50-002-33807 Supplier Medchemexpress LLC Supplier No. HY12160410MG
(Rac)-MEM 1003 is the racemate of MEM 1003, a dihydropyridine L-type calcium channel antagonist (CAS 165187-25-7) used in neuroscience research. The compound has formula C22H25ClN2O5 and molecular weight 432.9 g·mol⁻¹, and is supplied at high purity for research applications.
  • Racemic form of MEM 1003, a dihydropyridine calcium channel antagonist.
  • Potent L-type Ca2+ channel antagonist; useful in neuronal signaling studies.
  • High purity suitable for research use.
  • Available in small-scale quantities (e.g., 5 mg-100 mg) with larger sizes by request.
  • Certificate of analysis, datasheet, and safety data sheet available.