Learn More
Medchemexpress LLC Racemic triadimefon-d4 | MFCD00055506 | 99.5% | 297.77 g/mol | C14H12D4ClN3O2 | 5mg
SDP
Catalog No. 5000269126
Encompass_Preferred
( )-Triadimefon-d4 is deuterium labeled Triadimefon Triadimefon is a triazole fungicide used to control powdery mildew rusts and other fungal pests on grains fruit and vegetable crops turf shrubs and trees Triadimefon inhibits lanosterol 14 -demethylase interfering with oxidative demethylation reactions in the ergosterol biosynthesis pathway of fungi and also blocks gibberellin biosynthesis[1]
Catalog No. 50-002-69126 Supplier Medchemexpress LLC Supplier No. HYW7061805MG
( )-Triadimefon-d4 is deuterium labeled Triadimefon Triadimefon is a triazole fungicide used to control powdery mildew rusts and other fungal pests on grains fruit and vegetable crops turf shrubs and trees Triadimefon inhibits lanosterol 14 -demethylase interfering with oxidative demethylation reactions in the ergosterol biosynthesis pathway of fungi and also blocks gibberellin biosynthesis[1]