Description
- Scratch-resistant, clear polycarbonate lenses block >99.9% of harmful UV radiation
- Wraparound design offers superior side coverage
- Lightweight frames are durable and comfortable
- Flared single-lens design provides sleek protection
- A soft nose bridge and soft, bayonet-style side arms grip the face and temples lightly but firmly for a comfortable fit
- Choose from black or red nylon frames
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Frame Color | Red |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |