Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COGITA SCIENCE LLC RACK 16X100 OR 125MM TUBES

Catalog No. NC2249882
Encompass

Rack, Code 207. Holds 75 16x100 or 125mm Tubes

This item is not returnable. View return policy
Catalog No. NC2249882 Supplier COGITA SCIENCE LLC Supplier No. GILTR2504607
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rack, Code 207. Holds 75 16x100 or 125mm Tubes

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.