Learn More
Chemglass Life Sciences Rack, 5mm, NMR, Tubes, Sample, 72 Position
SDP
Catalog No. 50872786
Encompass_Preferred
Rack, 5mm, NMR, Tubes, Sample, 72 Position. These sturdy racks made from polypropylene have an upper deck with holes and a lower deck with indentations to support 5mm NMR tube ends. The sides have carrying handles. Dimensions are 21cm Wide x 11cm Deep x 22cm High.
Catalog No. 50-872-786 Supplier Chemglass Life Sciences Supplier No. CTR500
Rack, 5mm, NMR, Tubes, Sample, 72 Position. These sturdy racks made from polypropylene have an upper deck with holes and a lower deck with indentations to support 5mm NMR tube ends. The sides have carrying handles. Dimensions are 21cm Wide x 11cm Deep x 22cm High.