Learn More
MTC Bio Test Tube Rack, 60-Place (5 rows of 12), for centrifuge tubes, test tubes, culture tubes, and FACS tubes, 15-17mm diameter, PP, autoclavable, 245x110x72mm (L x W x H)
Catalog No. NC3737133
Encompass_Preferred
These autoclavable test tube racks are ideal for a variety of widely used laboratory tubes and vials. Manufactured in two sizes, these racks accommodate tubes ranging from 12-17mm in diameter, including test tubes, culture tubes, FACS tubes, large cryogenic vials, transport vials and more. Shipped in an economical flat pack, these racks snap together to create a sturdy and durable workstation. Molded alphanumeric grids along the sides assist in easy organization of samples. Available in 60 or 90 place, these heavy-duty polypropylene racks are autoclavable and can be safely used in water baths without floating.
Catalog No. NC3737133 Supplier MTC Bio Supplier No. R1117
These autoclavable test tube racks are ideal for a variety of widely used laboratory tubes and vials. Manufactured in two sizes, these racks accommodate tubes ranging from 12-17mm in diameter, including test tubes, culture tubes, FACS tubes, large cryogenic vials, transport vials and more. Shipped in an economical flat pack, these racks snap together to create a sturdy and durable workstation. Molded alphanumeric grids along the sides assist in easy organization of samples. Available in 60 or 90 place, these heavy-duty polypropylene racks are autoclavable and can be safely used in water baths without floating.