Learn More
Marlin Steel Wire Products Test Tube Rack 10.92"L x 5.59"W x 3"H
SDP
Catalog No. NC1810886
Encompass_Preferred
Quality engineered test tube rack used for sterilization, cleaning, and research. Holds 72 test tubes that are 18 mm (0.709") diameter tubesTypically used for autoclaving. *this item is custom and non returnable
Catalog No. NC1810886 Supplier Marlin Steel Wire Products Supplier No. 0068905534
Add to Cart
Add to Cart
Quality engineered test tube rack used for sterilization, cleaning, and research. Holds 72 test tubes that are 18 mm (0.709") diameter tubesTypically used for autoclaving. *this item is custom and non returnable