Learn More
Electron Microscopy Sciences Micro Sample Tube Rack with 8 Holes 10.8 mm each, 66 mm
SDP
Catalog No. 501899857
Encompass_Preferred
These round floating bubble racks can incubate 8 to 20 tubes in a beaker of water. The racks are loaded from the top and the tubes are easily removed by pressing the rack down on a bench until the short legs touch. Perfect for use in microbiology and immunology labs.
Molded from buoyant foam-filled TPXR (polymethylpentene) with tiny bubbles throughout. 10.8 mm diameter holes fit the 1.5 ml microtubes, and 7.7 mm holes fit the 0.5 ml tube. The racks are 1/4"(6.4 mm) thick with 3/4" (19 mm) long legs and a center rod handle. They are steam autoclavable at 250 degree F (121 degree C). Use in 400ml beakers. 10.8mm diameter holes fit the 1.5ml microtubes.
Hole: 10.8 mm
O.D.: 66 mm
Use: 400 mL beaker
Catalog No. 50-189-9857 Supplier Electron Microscopy Sciences Supplier No. 7236410
These round floating bubble racks can incubate 8 to 20 tubes in a beaker of water. The racks are loaded from the top and the tubes are easily removed by pressing the rack down on a bench until the short legs touch. Perfect for use in microbiology and immunology labs.
Molded from buoyant foam-filled TPXR (polymethylpentene) with tiny bubbles throughout. 10.8 mm diameter holes fit the 1.5 ml microtubes, and 7.7 mm holes fit the 0.5 ml tube. The racks are 1/4"(6.4 mm) thick with 3/4" (19 mm) long legs and a center rod handle. They are steam autoclavable at 250 degree F (121 degree C). Use in 400ml beakers. 10.8mm diameter holes fit the 1.5ml microtubes.
Hole: 10.8 mm
O.D.: 66 mm
Use: 400 mL beaker