Learn More
Azenta US Inc RACK 96 FORMAT 0.9ML W/CAPS 10
Supplier:  Azenta US Inc 6651016
Encompass
Rack, 96-format, HighBase, with TubeLock, 10 racks per case, suitable for 0.9ml External Thread Tubes with Screw Caps or Septum TPE Caps, 0.7ml Internal Thread Tubes with Screw Caps or Septum TPE Caps and 0.9ml Internal Thread Tubes with Septum TPE Caps
Catalog No. NC2284453