Learn More
Azenta US Inc RACK 96-FORMAT LOWBASE
Catalog No. NC2313495
Encompass
Rack, 96-format, LowBase, 10 racks per case, 2D rack ID position H12, suitable for 0.26ml Tubes with External Screw Caps
Catalog No. NC2313495 Supplier Azenta US Inc Supplier No. 68030020
Rack, 96-format, LowBase, 10 racks per case, 2D rack ID position H12, suitable for 0.26ml Tubes with External Screw Caps