Learn More
TrippNT 48 inch Adjustable Surgical Helmet Rack protects helmets like Stryker from damage and contamination. It has 5 adjustable hooks, each with a 50 lb capacity, and can hold other equipment. Made from corrosion-resistant HDPE.
SDP
Catalog No. NC3356643 Shop All TrippNT Products
Encompass_Preferred
TrippNT's 48 inch Adjustable Surgical Helmet Rack stores and protects surgical helmets from damage and contamination from falling. Designed in conjunction with a surgical department, this storage rack boasts 5 totally adjustable hooks giving you freedom to optimize placement of your helmets (styrker or similar). Also, a great option for hanging other oddly shaped equipment and supplies. Each hook has a 50 pound capacity. Constructed from non-metal white high-density polyethylene (HDPE), this product is corrosion resistant and easily sterilized. This rack ships with 3 wall anchors and screws for wall mounting. A smaller 24 inch (3 hook) size is available as well as individual replacement hooks. All TrippNT products are made in the USA and come with a 100% Total Replacement Lifetime Guarantee.
Catalog No. NC3356643 Supplier TrippNT Supplier No. 53940
TrippNT's 48 inch Adjustable Surgical Helmet Rack stores and protects surgical helmets from damage and contamination from falling. Designed in conjunction with a surgical department, this storage rack boasts 5 totally adjustable hooks giving you freedom to optimize placement of your helmets (styrker or similar). Also, a great option for hanging other oddly shaped equipment and supplies. Each hook has a 50 pound capacity. Constructed from non-metal white high-density polyethylene (HDPE), this product is corrosion resistant and easily sterilized. This rack ships with 3 wall anchors and screws for wall mounting. A smaller 24 inch (3 hook) size is available as well as individual replacement hooks. All TrippNT products are made in the USA and come with a 100% Total Replacement Lifetime Guarantee.