Learn More
Hach Company Bottle Rack, Plastic-Coated, 20 Compartments
Supplier:  Hach Company 2551801
Encompass
Made in the U.S.A. of 10 and 12-gauge steel fused together to create strong joints and straight lines. Rack is finished with an electrostatically applied powder coating. The result is a strong, durable finish which will stand up to many laboratory materials and the rack can be autoclaved. Max dry temperature is 300°F (148°C) and wet temperature is 500°F (260°C). Dimensions listed above are approximate. Compartments fit most bottles associated with standard drinking water sampling.Rack Size: 13-3/8" x 10-5/8" x 4" (34 x 27 x 10.2 cm)Compartment Size: 2-1/2" x 2-1/2" (6.4 x 6.4 cm)
Catalog No. 50-199-6217