Learn More
Beckman Coulter Beckman Coulter bucket holder rack for SW 32 Ti, SW 28.1 Ti, SW 28 rotors (P/N 331186)
Catalog No. NC2946427 Shop All Beckman Coulter Products
Encompass
Beckman Coulter bucket holder rack (P/N 331186) is a replacement accessory designed to secure and position swinging rotor buckets in compatible ultracentrifuge rotors. It is used for maintenance, rotor assembly, and as a spare part for swing-bucket rotor systems.
  • Holds swinging buckets securely during centrifugation.
  • Replacement rack for swing-bucket rotor assemblies.
  • Used for routine maintenance and rotor repair.
  • Compatible with commonly used swing-bucket rotor families.
Catalog No. NC2946427 Supplier Beckman Coulter Supplier No. 331186
Beckman Coulter bucket holder rack (P/N 331186) is a replacement accessory designed to secure and position swinging rotor buckets in compatible ultracentrifuge rotors. It is used for maintenance, rotor assembly, and as a spare part for swing-bucket rotor systems.
  • Holds swinging buckets securely during centrifugation.
  • Replacement rack for swing-bucket rotor assemblies.
  • Used for routine maintenance and rotor repair.
  • Compatible with commonly used swing-bucket rotor families.