Learn More
Hach Company Cooling rack for 10 cuvettes (Ø 16 mm), stainless steel
Supplier:  Hach Company 1864100
Encompass
Stainless steel rack for holding hot COD vials or test tubes prior to reading. Fit Hach COD vials and Test 'N Tube vials. Vials not included.
Catalog No. 50-199-5173