Learn More
Greiner Bio-One Rack with Cryo.s Biobanking Tubes and Capping Tool
GREENER_CHOICE
Catalog No. 07000975 Shop All Greiner Bio-One Products
Change view
Click to view available options
Capacity (Metric):
300 μL
600 μL
1000 μL
Packaging:
2 Racks/Bag
5 Racks/Bag
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Packaging
07-000-975 300 μL 5 Racks/Bag
07-000-976 600 μL 2 Racks/Bag
07-000-977 1000 μL 2 Racks/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-975 Supplier Greiner Bio-One Supplier No. 976561
Add to Cart
Add to Cart
Optimized for automated laboratory equipment such as decappers, liquid handling systems and automated storage systems.
Rack:
  • Stackable, with footprint
  • Rack bottom with scanning windows for Datamatrix, rotation stoppers, snap-in lid
Tubes:
  • Fitted with screw caps (made of silicone gasket and inte
Compliance
Corresponds to American National Standards Institute (ANSI-Standard)
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Capacity (Metric) 300 μL
Material Polycarbonate
Packaging 5 Racks/Bag
RNase Free Yes
Sterility Sterile
Graduated Yes
Closure Type Silicone Gasket, Internal Thread